Assay Details
| Gene Symbol: acvr1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130990.1 | NP_571065.1 | 3 - 4 | 576 | 90 | |
| XM_005162224.4 | XP_005162281.1 | 3 - 4 | 90 | |||
| GenBank mRNA | BC163346.1 | - | 3 - 4 | 90 | ||
| BC163377.1 | - | 3 - 4 | 90 | |||
| X94119.1 | - | 576 | 90 | |||
More Information
Set Membership: |
|

acvr1ba
Genomic Map