Assay Details
| Gene Symbol: sema3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131061.1 | NP_571136.1 | 4 - 5 | 454 | 60 | |
| GenBank mRNA | AF083382.1 | - | 4 - 5 | 454 | 60 | |
| BC163261.1 | - | 4 - 5 | 534 | 60 | ||
| CU458856.1 | - | 4 - 5 | 454 | 60 | ||
More Information
Set Membership: |
|

sema3ab
Genomic Map