Assay Details
| Gene Symbol: nr1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131065.1 | NP_571140.1 | 2 - 3 | 637 | 81 | |
| GenBank mRNA | AF342942.1 | - | 2 - 3 | 269 | 81 | |
| BC074065.1 | - | 2 - 3 | 637 | 81 | ||
| BC164697.1 | - | 2 - 3 | 261 | 81 | ||
More Information
Set Membership: |
|

nr1d2b
Genomic Map