Assay Details
| Gene Symbol: nr1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131065.1 | NP_571140.1 | 5 - 6 | 1552 | 88 | |
| XM_005159500.4 | XP_005159557.1 | 4 - 5 | 1485 | 88 | ||
| GenBank mRNA | AF342942.1 | - | 5 - 6 | 1184 | 88 | |
| BC074065.1 | - | 5 - 6 | 1552 | 88 | ||
| BC164697.1 | - | 5 - 6 | 1176 | 88 | ||
More Information
Set Membership: |
|

nr1d2b
Genomic Map