Assay Details
Gene Symbol: apoeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131098.2 | NP_571173.1 | 4 - 5 | 686 | 65 | |
GenBank mRNA | BC049470.1 | - | 4 - 5 | 533 | 65 | |
BC065592.1 | - | 3 - 4 | 519 | 65 | ||
BC154034.1 | - | 4 - 5 | 535 | 65 | ||
Y13652.1 | - | 493 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
