Assay Details
Gene Symbol: nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131145.2 | NP_571220.2 | 4 - 5 | 743 | 91 | |
GenBank mRNA | BC076419.1 | - | 4 - 5 | 722 | 91 | |
EH537868.1 | - | 4 - 5 | 782 | 91 | ||
U62619.1 | - | 4 - 5 | 732 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
