Assay Details
Gene Symbol: pax7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131325.2 | NP_571400.1 | 3 - 4 | 806 | 71 | |
NM_131332.2 | NP_571407.2 | 3 - 4 | 806 | 71 | ||
GenBank mRNA | AF014367.1 | - | 3 - 4 | 807 | 71 | |
AF014368.1 | - | 3 - 4 | 807 | 71 | ||
AF014369.1 | - | 3 - 4 | 807 | 71 | ||
BC163492.1 | - | 3 - 4 | 575 | 71 | ||
BC163502.1 | - | 3 - 4 | 575 | 71 | ||
BC163523.1 | - | 3 - 4 | 575 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
