Assay Details
| Gene Symbol: pax7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131325.2 | NP_571400.1 | 4 - 5 | 933 | 63 | |
| NM_131332.2 | NP_571407.2 | 4 - 5 | 933 | 63 | ||
| XM_009306286.3 | XP_009304561.1 | 4 - 5 | 1038 | 63 | ||
| GenBank mRNA | AF014367.1 | - | 4 - 5 | 934 | 63 | |
| AF014368.1 | - | 4 - 5 | 934 | 63 | ||
| AF014369.1 | - | 4 - 5 | 934 | 63 | ||
| BC163492.1 | - | 4 - 5 | 702 | 63 | ||
| BC163502.1 | - | 4 - 5 | 702 | 63 | ||
| BC163523.1 | - | 4 - 5 | 702 | 63 | ||
| BC163580.1 | - | 4 - 5 | 687 | 63 | ||
More Information
Set Membership: |
|

pax7a
Genomic Map