Assay Details
| Gene Symbol: mid1ip1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131335.1 | NP_571410.1 | 1 | 452 | 83 | |
| GenBank mRNA | BC074084.1 | - | 1 | 428 | 83 | |
| BC164900.1 | - | 1 | 423 | 83 | ||
| U27121.1 | - | 1 | 452 | 83 | ||
More Information
Set Membership: |
|

mid1ip1a
Genomic Map