Assay Details
| Gene Symbol: stat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131480.1 | NP_571555.1 | 21 - 22 | 2148 | 81 | |
| GenBank mRNA | AJ005692.1 | - | 21 - 22 | 2148 | 81 | |
| BC045863.1 | - | 20 - 21 | 2295 | 81 | ||
| BC060933.1 | - | 21 - 22 | 2177 | 81 | ||
More Information
Set Membership: |
|

stat1a
Genomic Map