Assay Details
| Gene Symbol: stat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131480.1 | NP_571555.1 | 6 - 7 | 641 | 120 | |
| GenBank mRNA | AJ005692.1 | - | 6 - 7 | 641 | 120 | |
| BC045863.1 | - | 6 - 7 | 699 | 120 | ||
| BC060933.1 | - | 6 - 7 | 670 | 120 | ||
More Information
Set Membership: |
|

stat1a
Genomic Map