Assay Details
Gene Symbol: gch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131667.1 | NP_571742.1 | 4 - 5 | 633 | 84 | |
GenBank mRNA | AJ311846.1 | - | 4 - 5 | 621 | 84 | |
BC071298.1 | - | 4 - 5 | 633 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
