Assay Details
| Gene Symbol: angptl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131818.3 | NP_571893.2 | 6 - 7 | 1325 | 73 | |
| XM_021476750.1 | XP_021332425.1 | 7 - 8 | 1805 | 73 | ||
| XM_021476751.1 | XP_021332426.1 | 7 - 8 | 1758 | 73 | ||
| GenBank mRNA | AF379604.2 | - | 6 - 7 | 1220 | 73 | |
| BC163088.1 | - | 6 - 7 | 1281 | 73 | ||
More Information
Set Membership: |
|

angptl3
Genomic Map