Assay Details
Gene Symbol: pcdh10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182885.1 | NP_878305.1 | 2 - 3 | 3008 | 67 | |
XM_009291096.3 | XP_009289371.1 | 2 - 3 | 3053 | 67 | ||
GenBank mRNA | AB086625.1 | - | 2 - 3 | 3008 | 67 | |
BC070019.1 | - | 2 - 3 | 2608 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
