Assay Details
| Gene Symbol: mcl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194394.2 | NP_919375.2 | 1 - 2 | 450 | 106 | |
| GenBank mRNA | AF441284.1 | - | 1 - 2 | 326 | 106 | |
| BC092680.1 | - | 1 - 2 | 450 | 106 | ||
| BC165058.1 | - | 1 - 2 | 360 | 106 | ||
More Information
Set Membership: |
|

mcl1b
Genomic Map