Assay Details
| Gene Symbol: mcl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194394.2 | NP_919375.2 | 2 - 3 | 700 | 70 | |
| GenBank mRNA | AF441284.1 | - | 2 - 3 | 576 | 70 | |
| BC092680.1 | - | 2 - 3 | 700 | 70 | ||
| BC165058.1 | - | 2 - 3 | 610 | 70 | ||
More Information
Set Membership: |
|

mcl1b
Genomic Map