Assay Details
| Gene Symbol: p2rx3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198986.3 | NP_945337.3 | 2 - 3 | 456 | 73 | |
| XM_021466489.1 | XP_021322164.1 | 2 - 3 | 522 | 73 | ||
| GenBank mRNA | AY292651.1 | - | 2 - 3 | 285 | 73 | |
| BC095293.1 | - | 2 - 3 | 457 | 73 | ||
More Information
Set Membership: |
|

p2rx3b
Genomic Map