Assay Details
Gene Symbol: p2rx3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198986.3 | NP_945337.3 | 7 - 8 | 911 | 77 | |
XM_021466489.1 | XP_021322164.1 | 7 - 8 | 977 | 77 | ||
GenBank mRNA | AY292651.1 | - | 7 - 8 | 740 | 77 | |
BC095293.1 | - | 7 - 8 | 912 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
