Assay Details
Gene Symbol: evla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199519.3 | NP_955813.3 | 12 - 13 | 1524 | 76 | |
XM_005158792.4 | XP_005158849.1 | 13 - 14 | 2150 | 76 | ||
XM_005158793.4 | XP_005158850.1 | 13 - 14 | 2143 | 76 | ||
XM_005158794.4 | XP_005158851.1 | 13 - 14 | 1938 | 76 | ||
XM_005158795.4 | XP_005158852.1 | 13 - 14 | 2277 | 76 | ||
XM_005158796.4 | XP_005158853.1 | 11 - 12 | 1854 | 76 | ||
XM_005158797.4 | XP_005158854.1 | 12 - 13 | 1391 | 76 | ||
XM_005158798.4 | XP_005158855.1 | 12 - 13 | 1389 | 76 | ||
XM_005158799.4 | XP_005158856.1 | 12 - 13 | 1386 | 76 | ||
XM_021467223.1 | XP_021322898.1 | 13 - 14 | 2148 | 76 | ||
GenBank mRNA | BC056325.1 | - | 12 - 13 | 1495 | 76 | |
BC065353.1 | - | 12 - 13 | 1488 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
