Assay Details
Gene Symbol: evla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199519.3 | NP_955813.3 | 9 - 10 | 1338 | 105 | |
XM_005158792.4 | XP_005158849.1 | 10 - 11 | 1964 | 105 | ||
XM_005158793.4 | XP_005158850.1 | 10 - 11 | 1957 | 105 | ||
XM_005158794.4 | XP_005158851.1 | 10 - 11 | 1752 | 105 | ||
XM_005158795.4 | XP_005158852.1 | 10 - 11 | 2091 | 105 | ||
XM_005158796.4 | XP_005158853.1 | 8 - 9 | 1668 | 105 | ||
XM_005158797.4 | XP_005158854.1 | 9 - 10 | 1205 | 105 | ||
XM_005158798.4 | XP_005158855.1 | 9 - 10 | 1203 | 105 | ||
XM_005158799.4 | XP_005158856.1 | 9 - 10 | 1200 | 105 | ||
XM_021467223.1 | XP_021322898.1 | 10 - 11 | 1962 | 105 | ||
GenBank mRNA | BC056325.1 | - | 9 - 10 | 1309 | 105 | |
BC065353.1 | - | 9 - 10 | 1302 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
