Assay Details
Gene Symbol: setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199600.1 | NP_955894.1 | 2 - 3 | 276 | 81 | |
GenBank mRNA | BC044440.1 | - | 2 - 3 | 292 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
