Assay Details
Gene Symbol: cbx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199746.2 | NP_956040.2 | 3 - 4 | 488 | 67 | |
GenBank mRNA | BC045443.1 | - | 3 - 4 | 474 | 67 | |
BC075782.1 | - | 3 - 4 | 436 | 67 | ||
BC165019.1 | - | 2 - 3 | 253 | 67 | ||
EH457628.1 | - | 3 - 4 | 494 | 67 | ||
EH545424.1 | - | 3 - 4 | 527 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
