Assay Details
| Gene Symbol: znf710a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199792.1 | NP_956086.1 | 3 - 4 | 1808 | 76 | |
| XM_005159166.4 | XP_005159223.1 | 3 - 4 | 2367 | 76 | ||
| XM_005159167.4 | XP_005159224.1 | 3 - 4 | 2337 | 76 | ||
| GenBank mRNA | BC054704.1 | - | 3 - 4 | 1808 | 76 | |
More Information
Set Membership: |
|

znf710a
Genomic Map