Assay Details
Gene Symbol: larp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199903.1 | NP_956197.1 | 2 - 3 | 565 | 81 | |
GenBank mRNA | BC056595.1 | - | 2 - 3 | 565 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
