Assay Details
Gene Symbol: wasb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199938.1 | NP_956232.1 | 4 - 5 | 542 | 85 | |
GenBank mRNA | BC053291.1 | - | 4 - 5 | 542 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
