Assay Details
Gene Symbol: wasb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199938.1 | NP_956232.1 | 5 - 6 | 574 | 81 | |
GenBank mRNA | BC053291.1 | - | 5 - 6 | 574 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
