Assay Details
Gene Symbol: chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200057.1 | NP_956351.1 | 2 - 3 | 287 | 62 | |
GenBank mRNA | BC054696.1 | - | 2 - 3 | 287 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
