Assay Details
| Gene Symbol: reep3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200161.1 | NP_956455.1 | 2 - 3 | 369 | 87 | |
| GenBank mRNA | BC045373.1 | - | 2 - 3 | 369 | 87 | |
More Information
Set Membership: |
|

reep3b
Genomic Map