Assay Details
Gene Symbol: reep3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200161.1 | NP_956455.1 | 6 - 7 | 817 | 68 | |
GenBank mRNA | BC045373.1 | - | 6 - 7 | 817 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
