Assay Details
| Gene Symbol: mboat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200537.1 | NP_956831.1 | 2 - 3 | 155 | 124 | |
| XM_021466647.1 | XP_021322322.1 | 2 - 3 | 250 | 124 | ||
| GenBank mRNA | BC056305.1 | - | 2 - 3 | 155 | 124 | |
More Information
Set Membership: |
|

mboat7
Genomic Map