Assay Details
| Gene Symbol: tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348480.1 | NP_001335409.1 | 2 - 3 | 648 | 78 | |
| NM_200569.2 | NP_956863.1 | 2 - 3 | 343 | 78 | ||
| GenBank mRNA | BC056586.1 | - | 2 - 3 | 331 | 78 | |
| BC068985.1 | - | 2 - 3 | 327 | 78 | ||
More Information
Set Membership: |
|

tsc22d3
Genomic Map