Assay Details
| Gene Symbol: cacng2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200641.1 | NP_956935.1 | 2 - 3 | 786 | 149 | |
| GenBank mRNA | BC057439.1 | - | 2 - 3 | 786 | 149 | |
More Information
Set Membership: |
|

cacng2a
Genomic Map