Assay Details
| Gene Symbol: rheb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200729.1 | NP_957023.1 | 3 - 4 | 520 | 128 | |
| GenBank mRNA | BC059505.1 | - | 3 - 4 | 520 | 128 | |
| BC163610.1 | - | 3 - 4 | 432 | 128 | ||
More Information
Set Membership: |
|

rheb
Genomic Map