Assay Details
Gene Symbol: klc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200953.2 | NP_957247.1 | 13 - 14 | 1420 | 66 | |
XM_005156633.4 | XP_005156690.1 | 13 - 14 | 1908 | 66 | ||
XM_005156635.4 | XP_005156692.1 | 13 - 14 | 1857 | 66 | ||
XM_005156637.3 | XP_005156694.1 | 13 - 14 | 1857 | 66 | ||
XM_005156638.3 | XP_005156695.1 | 13 - 14 | 1857 | 66 | ||
XM_009307227.3 | XP_009305502.1 | 13 - 14 | 1857 | 66 | ||
GenBank mRNA | BC055591.1 | - | 13 - 14 | 1421 | 66 | |
BC165713.1 | - | 12 - 13 | 1380 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
