Assay Details
Gene Symbol: klc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200953.2 | NP_957247.1 | 2 - 3 | 351 | 144 | |
GenBank mRNA | BC055591.1 | - | 2 - 3 | 352 | 144 | |
BC165713.1 | - | 1 - 2 | 311 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
