Assay Details
Gene Symbol: klc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200953.2 | NP_957247.1 | 9 - 10 | 1090 | 68 | |
XM_005156633.4 | XP_005156690.1 | 9 - 10 | 1578 | 68 | ||
XM_005156635.4 | XP_005156692.1 | 9 - 10 | 1527 | 68 | ||
XM_005156637.3 | XP_005156694.1 | 9 - 10 | 1527 | 68 | ||
XM_005156638.3 | XP_005156695.1 | 9 - 10 | 1527 | 68 | ||
XM_009307227.3 | XP_009305502.1 | 9 - 10 | 1527 | 68 | ||
GenBank mRNA | BC055591.1 | - | 9 - 10 | 1091 | 68 | |
BC165713.1 | - | 8 - 9 | 1050 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
