Assay Details
| Gene Symbol: sh2d3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201076.1 | NP_957370.1 | 2 - 3 | 259 | 69 | |
| GenBank mRNA | BC049321.1 | - | 2 - 3 | 259 | 69 | |
More Information
Set Membership: |
|

sh2d3cb
Genomic Map