Assay Details
| Gene Symbol: sh2d3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201076.1 | NP_957370.1 | 9 - 10 | 1771 | 150 | |
| XM_005165604.4 | XP_005165661.1 | 9 - 10 | 1708 | 150 | ||
| XM_021476041.1 | XP_021331716.1 | 9 - 10 | 1693 | 150 | ||
| GenBank mRNA | BC049321.1 | - | 9 - 10 | 1771 | 150 | |
More Information
Set Membership: |
|

sh2d3cb
Genomic Map