Assay Details
Gene Symbol: dcun1d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205538.1 | NP_991101.1 | 4 - 5 | 539 | 85 | |
XM_005167810.4 | XP_005167867.1 | 4 - 5 | 644 | 85 | ||
XM_005167811.4 | XP_005167868.1 | 4 - 5 | 666 | 85 | ||
XM_021478598.1 | XP_021334273.1 | 4 - 5 | 611 | 85 | ||
GenBank mRNA | BC065884.1 | - | 4 - 5 | 539 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
