Assay Details
Gene Symbol: pnp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205655.2 | NP_991218.2 | 3 - 4 | 601 | 119 | |
GenBank mRNA | BC065621.1 | - | 3 - 4 | 557 | 119 | |
CK028065.1 | - | 3 - 4 | 432 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
