Assay Details
| Gene Symbol: etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212998.2 | NP_998163.1 | 5 - 6 | 608 | 72 | |
| GenBank mRNA | BC045971.1 | - | 5 - 6 | 609 | 72 | |
| BC164332.1 | - | 5 - 6 | 626 | 72 | ||
More Information
Set Membership: |
|

etfb
Genomic Map