Assay Details
| Gene Symbol: asic4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214787.2 | NP_999952.1 | 7 - 8 | 1539 | 132 | |
| GenBank mRNA | AJ609619.1 | - | 7 - 8 | 1540 | 132 | |
More Information
Set Membership: |
|

asic4a
Genomic Map