Assay Details
| Gene Symbol: rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002160.2 | NP_001002160.2 | 356 | 149 | ||
| NM_001244839.1 | NP_001231768.1 | 2 - 3 | 347 | 140 | ||
| GenBank mRNA | BC071307.1 | - | 149 | |||
| EH455622.1 | - | 382 | 149 | |||
| EH536597.1 | - | 2 - 3 | 390 | 140 | ||
More Information
Set Membership: |
|

rpf1
Genomic Map