Assay Details
Gene Symbol: ldhbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002474.1 | NP_001002474.1 | 3 - 4 | 407 | 82 | |
XM_005170658.4 | XP_005170715.1 | 3 - 4 | 515 | 82 | ||
XM_005170659.2 | XP_005170716.1 | 3 - 4 | 411 | 82 | ||
XM_021470477.1 | XP_021326152.1 | 3 - 4 | 511 | 82 | ||
GenBank mRNA | BC076340.1 | - | 3 - 4 | 407 | 82 | |
BC165286.1 | - | 2 - 3 | 281 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
