Assay Details
Gene Symbol: spryd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002477.1 | NP_001002477.1 | 1 - 2 | 158 | 146 | |
GenBank mRNA | BC076335.1 | - | 1 - 2 | 158 | 146 | |
BC152179.1 | - | 1 - 2 | 199 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
