Assay Details
| Gene Symbol: mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002567.1 | NP_001002567.1 | 1 - 2 | 536 | 59 | |
| GenBank mRNA | BC076231.1 | - | 1 - 2 | 536 | 59 | |
| BC086824.1 | - | 1 - 2 | 515 | 59 | ||
More Information
Set Membership: |
|

mpv17l2
Genomic Map