Assay Details
| Gene Symbol: suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002577.2 | NP_001002577.2 | 4 - 5 | 504 | 61 | |
| GenBank mRNA | BC076216.1 | - | 4 - 5 | 492 | 61 | |
| BC081520.1 | - | 4 - 5 | 504 | 61 | ||
| BC164126.1 | - | 4 - 5 | 504 | 61 | ||
More Information
Set Membership: |
|

suclg1
Genomic Map