Assay Details
| Gene Symbol: faah2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002700.2 | NP_001002700.1 | 2 - 3 | 325 | 142 | |
| GenBank mRNA | BC076113.1 | - | 2 - 3 | 326 | 142 | |
More Information
Set Membership: |
|

faah2a
Genomic Map