Assay Details
| Gene Symbol: ndufab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003418.1 | NP_001003418.1 | 3 - 4 | 409 | 88 | |
| GenBank mRNA | BC076098.1 | - | 3 - 4 | 409 | 88 | |
More Information
Set Membership: |
|

ndufab1a
Genomic Map