Assay Details
| Gene Symbol: mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003439.1 | NP_001003439.1 | 7 - 8 | 575 | 69 | |
| GenBank mRNA | BC076155.1 | - | 7 - 8 | 575 | 69 | |
| BC153656.1 | - | 7 - 8 | 579 | 69 | ||
More Information
Set Membership: |
|

mob4
Genomic Map